accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOO

CAS RN 745783-88-4

IUPAC Name: 2-(6-methoxynaphthalen-2-yl)-2-oxoacetaldehyde hydrate
Synonyms: 2-(6-methoxynaphthalen-2-yl)-2-oxoacetaldehyde hydrate
Molecular Weight (g/mol): 232.24
Molecular Formula: C13H12O4
InChi Key: YNILJUBMHVAHAW-UHFFFAOYSA-N
SMILES: O.COC1=CC=C2C=C(C=CC2=C1)C(=O)C=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)