CAS RN 7472-43-7
IUPAC Name:
7-oxo-7-phenylheptanoic acid
Synonyms:
7-oxo-7-phenylheptanoic acid
Molecular Weight (g/mol):
220.27
Molecular Formula:
C13H16O3
InChi Key:
DOQWHEUDAHLEPT-UHFFFAOYSA-N
SMILES:
OC(=O)CCCCCC(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
6-Benzoylhexanoic acid, 94%, Thermo Scientific Chemicals
CAS: 7472-43-7 Molecular Formula: C13H16O3 Molecular Weight (g/mol): 220.27 MDL Number: MFCD00039608 InChI Key: DOQWHEUDAHLEPT-UHFFFAOYSA-N Synonym: 6-benzoylhexanoic acid,7-oxo-7-phenyl-heptanoic acid,6-benzoyl hexanoic acid,benzeneheptanoic acid, z-oxo,7-oxo-7-phenylheptanoicacid,#,benzeneheptanoic acid,,ae-oxo,alpha-oxo benzeneheptanoic acid,benzeneheptanoic acid,alpha-oxo PubChem CID: 344830 IUPAC Name: 7-oxo-7-phenylheptanoic acid SMILES: OC(=O)CCCCCC(=O)C1=CC=CC=C1