accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 75-36-5

IUPAC Name: acetyl chloride
Synonyms: acetyl chloride acetic chloride acetyl chloride (substance) C2H3ClO rcra waste number U006
Molecular Weight (g/mol): 78.5
Molecular Formula: C2H3ClO
InChi Key: WETWJCDKMRHUPV-UHFFFAOYSA-N
SMILES: CC(Cl)=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (16)
Boiling Point 
  • (6)
  • (3)
  • (6)
Physical Form 
  • (12)
Percent Purity 
  • (1)
  • (5)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (16)
  • (6)
  • (3)
  • (6)
  • (12)
  • (1)
  • (5)
  • (4)