CAS RN 75-65-0
tert-Butanol, 99.5%, extra pure
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butyl alcohol, anhydrous, 99.5%
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butanol, ACS reagent
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
Tert-Butyl Alcohol, Certified AR for Analysis, Fisher Chemical™
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: 4464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butyl alcohol, 99%
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butanol, 99.5%, for analysis
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butyl alcohol, ACS, 99+%
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butanol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Tert-Butanol, puriss. p.a. ACS Reagent, ≥99.7% (GC), Solstice
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butanol, CHROMASOLV™, for HPLC, ≥99.5%, Solstice
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
Tert-Butyl Alcohol, Extra Pure, SLR, Fisher Chemical™
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: 4464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butanol, TEBOL™, ≥99.3%, Honeywell™
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butyl Alcohol, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
- 1
- 2