accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 75-85-4

H3CCH3CH3HO

CAS RN 75-85-4

IUPAC Name: 2-methylbutan-2-ol
Synonyms: 2-methyl-2-butanol dimethyl ethyl carbinol ethyl dimethyl carbinol tertiary amyl alcohol tertiaerer amylalkohol amylalkohol, tertiaerer tert-isoamyl alcohol amylenum hydratum potassium t-amylate
Molecular Weight (g/mol): 88.15
Molecular Formula: C5H12O
InChi Key: MSXVEPNJUHWQHW-UHFFFAOYSA-N
SMILES: CCC(C)(C)O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (8)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)