accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OBrBrF

CAS RN 7542-64-5

IUPAC Name: 2,2-dibromo-1-(4-fluorophenyl)ethan-1-one
Synonyms: 2,2-dibromo-1-(4-fluorophenyl)ethanone
Molecular Weight (g/mol): 295.93
Molecular Formula: C8H5Br2FO
InChi Key: XFFGHBAVJPOWDG-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)C(Br)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results