accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH(S)OOOOO

CAS RN 75507-25-4

IUPAC Name: [(2S)-1,4,7,10,13-pentaoxacyclopentadecan-2-yl]methanol
Synonyms: (2S)-1,4,7,10,13-pentaoxacyclopentadecan-2-ylmethanol
Molecular Weight (g/mol): 250.29
Molecular Formula: C11H22O6
InChi Key: YHIQMMGCRYKJLB-NSHDSACASA-N
SMILES: OC[C@H]1COCCOCCOCCOCCO1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)