accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3COO(S)(S)(R)H3C(R)(R)(S)HO(R)HO(S)HOOCH3CH3H3C(Z)

CAS RN 75567-37-2

IUPAC Name: (1S,4S,5S,6R,9S,10R,12R,14R)-5,6-dihydroxy-7-(hydroxymethyl)-3,11,11,14-tetramethyl-15-oxotetracyclo[7.5.1.0¹,⁵.0¹⁰,¹²]pentadeca-2,7-dien-4-yl (2Z)-2-methylbut-2-enoate
Synonyms: ingenol mebutate
Molecular Weight (g/mol): 430.54
Molecular Formula: C25H34O6
InChi Key: VDJHFHXMUKFKET-WDUFCVPESA-N
SMILES: C\C=C(\C)C(=O)O[C@H]1C(C)=C[C@@]23[C@H](C)C[C@@H]4[C@H]([C@H](C=C(CO)[C@@H](O)[C@]12O)C3=O)C4(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results