missing translation for 'onlineSavingsMsg'
Learn More

Ingenol 3-Angelate, TRC

Product Code. 30408627
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408627 10 mg -
30404954 2.5 mg 2.5mg
30395466 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408627 Supplier Toronto Research Chemicals Supplier No. TRCI65580010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75567-37-2
Molecular Formula C25 H34 O6
Molecular Weight (g/mol) 430.53
Synonym 2-Butenoic acid, 2-methyl-, (1aR,2S,5R,5aS,6S,8aS,9R,10aR)-1a,2,5,5a,6,9,10,10a-octahydro-5,5a-dihydroxy-4-(hydroxymethyl)-1,1,7,9-tetramethyl-11-oxo-1H-2,8a-methanocyclopenta[a]cyclopropa[e]cyclodecen-6-yl ester, (2Z)-, 2-Butenoic acid, 2-methyl-, 1a,2,5,5a,6,9,10,10a-octahydro-5,5a-dihydroxy-4-(hydroxymethyl)-1,1,7,9-tetramethyl-11-oxo-1H-2,8a-methanocyclopenta[a]cyclopropa[e]cyclodecen-6-yl ester, [1aR-[1aα,2β,5β,5aβ,6β(Z),8aα,9α,10aα]]-, 1H-2,8a-Methanocyclopenta[a]cyclopropa[e]cyclodecene, 2-butenoic acid deriv., 3-Angeloylingenol, 3-Ingenyl angelate, Euphorbia factor An1, Euphorbia factor H1, Ingenol 3-angelate, Ingenol 3-mebutate, Ingenol mebutate, PEP 005, Picato
SMILES C\C=C(\C)/C(=O)O[C@H]1C(=C[C@@]23[C@H](C)C[C@@H]4[C@H]([C@H](C=C(CO)[C@@H](O)[C@]12O)C3=O)C4(C)C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C25H34O6/c1-7-12(2)22(29)31-21-13(3)10-24-14(4)8-17-18(23(17,5)6)16(20(24)28)9-15(11-26)19(27)25(21,24)30/h7,9-10,14,16-19,21,26-27,30H,8,11H2,1-6H3/b12-7-/t14-,16+,17-,18+,19-,21+,24+,25+/m1/s1
Formula Weight 430.2355
Chemical Name or Material Ingenol Mebutate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.