accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH3CS

CAS RN 758-16-7

IUPAC Name: N,N-dimethylmethanethioamide
Synonyms: N,N-dimethylmethanethioamide
Molecular Weight (g/mol): 89.16
Molecular Formula: C3H7NS
InChi Key: SKECXRFZFFAANN-UHFFFAOYSA-N
SMILES: CN(C)C=S
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)