missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylthioformamide, 97%

Product Code. 11490358
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490358 25 g 25g
11480358 5 g 5g
11400368 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490358 Supplier Thermo Scientific Alfa Aesar Supplier No. B24529.14
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 758-16-7 | C3H7NS | 89.156 g/mol

It is used in telecommunication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in telecommunication.

Solubility
Fully miscible in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 758-16-7
Molecular Formula C3H7NS
Molecular Weight (g/mol) 89.156
MDL Number MFCD00004943
InChI Key SKECXRFZFFAANN-UHFFFAOYSA-N
Synonym n,n-dimethylthioformamide, dimethylthioformamide, methanethioamide, n,n-dimethyl, formamide, n,n-dimethylthio, nn-dimethylthioformamide, methanethioamide,n,n-dimethyl, formamide,n-dimethylthio, dimethylamino methanethial, n,n-dimethyl-thioformamide, methanethioamide, dimethyl
PubChem CID 69794
IUPAC Name N,N-dimethylmethanethioamide
SMILES CN(C)C=S

Specifications

Boiling Point 58°C to 60°C (1 mmHg)
Quantity 25 g
Solubility Information Fully miscible in water.
Formula Weight 89.15
Percent Purity 97%
Chemical Name or Material N,N-Dimethylthioformamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.