accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOCH3CH3H3CCH3

CAS RN 759-24-0

IUPAC Name: 1,3-diethyl 2-tert-butylpropanedioate
Synonyms: 1,3-diethyl 2-tert-butylpropanedioate
Molecular Weight (g/mol): 216.28
Molecular Formula: C11H20O4
InChi Key: RJNICNBRGVKNSR-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C(=O)OCC)C(C)(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results