accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 76093-98-6

IUPAC Name: pentacontabismuth dodecacadmium pentacosalead dodecatin
Synonyms: pentacontabismuth dodecacadmium pentacosalead dodecatin
Molecular Weight (g/mol): 18402.5
Molecular Formula: Bi50Cd12Pb25Sn12
InChi Key: CQHDPRBPWAYYKI-UHFFFAOYSA-N
SMILES: [Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Sn].[Sn].[Sn].[Sn].[Sn].[Sn].[Sn].[Sn].[Sn].[Sn].[Sn].[Sn].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Pb].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi].[Bi]
Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results