missing translation for 'onlineSavingsMsg'
Learn More

Wood's Metal, Pure, M.P.70°C

Product Code. 10647042 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10647042 500 g 500g
10142210 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10647042 Supplier Fisher Chemical Supplier No. W/0350/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 76093-98-6
Molecular Formula Bi·Cd·Pb·Sn
MDL Number 198175

Specifications

Color Gray
Melting Point 70°C
Quantity 500 g
Packaging Plastic powder jar
Form Solid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Wood's metal m.p. ca. 70°C
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H362-May cause harm to breast-fed children.
  • H372-Causes damage to organs.
  • H360FD-May damage fertility. May damage the unborn child.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P263-Avoid contact during pregnancy/while nursing.
Supplemental information
  • MIXTURE LIST-Contain: Bismuth alloy, base,Bi 50, Pb 25, Cd 12, Sn 12, Cadmium, Tin, Lead
  • REA17-CMR1-Restricted to professional users
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.