accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 762-04-9

H3COPOOCH3

CAS RN 762-04-9

IUPAC Name: diethoxy(oxo)-λ⁵-phosphanylium
Synonyms: diethoxy-oxophosphanium
Molecular Weight (g/mol): 137.09
Molecular Formula: C4H10O3P
InChi Key: LXCYSACZTOKNNS-UHFFFAOYSA-N
SMILES: CCO[P+](=O)OCC
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)