CAS RN 7647-15-6
Sodium bromide, ACS reagent
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Sodium bromide, 99+%, extra pure, anhydrous
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 IUPAC Name: sodium bromide SMILES: [Na+].[Br-]
Sodium bromide, 99.5%, for analysis
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Sodium bromide, 97%
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 IUPAC Name: sodium bromide SMILES: [Na+].[Br-]
Sodium bromide, 99+% (dry wt.), water <1.0%
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Sodium Bromide, Solstice
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Sodium Bromide, Extra Pure, SLR, Fisher Chemical™
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Sodium bromide, ACS, 99.0% min
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Sodium bromide, ultra dry, 99.999% (metals basis)
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 IUPAC Name: sodium bromide SMILES: [Na+].[Br-]
Sodium bromide, 99.99% (metals basis)
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Sodium bromide, Puratronic™, 99.9955% (metals basis)
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Thermo Scientific Chemicals Sodium bromide, 99+%, for biochemistry
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Sodium Bromide, ≥99.0%, ACS Reagent, Solstice
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 IUPAC Name: sodium;bromide SMILES: [Na+].[Br-]
Bromide Standard for Ion Chromatography, 0.1M NaBr, SPEX CertiPrep™
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 IUPAC Name: sodium bromide SMILES: [Na+].[Br-]
Bromide Standard for Ion Chromatography, 1000μg/mL Br-, SPEX CertiPrep™
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 IUPAC Name: sodium bromide SMILES: [Na+].[Br-]
- 1
- 2