accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

INO

CAS RN 766557-62-4

IUPAC Name: 2-(cyclopropylmethoxy)-3-iodopyridine
Synonyms: 2-(cyclopropylmethoxy)-3-iodopyridine
Molecular Weight (g/mol): 275.09
Molecular Formula: C9H10INO
InChi Key: ILSBVXOYWKUWLJ-UHFFFAOYSA-N
SMILES: IC1=CC=CN=C1OCC1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results