accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSCH3NH3C

CAS RN 76788-46-0

IUPAC Name: 2-ethyl-4,5-dimethyl-2,5-dihydro-1,3-thiazole
Synonyms: 2-ethyl-4,5-dimethyl-2,5-dihydro-1,3-thiazole
Molecular Weight (g/mol): 143.25
Molecular Formula: C7H13NS
InChi Key: CSACPVARWHDAET-UHFFFAOYNA-N
SMILES: CCC1SC(C)C(C)=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results