missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-4,5-dimethyl-3-thiazoline, cis + trans, 99%, Thermo Scientific™

Product Code. 11431570
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431570 10 g 10g
11421570 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431570 Supplier Thermo Scientific Alfa Aesar Supplier No. H26960.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76788-46-0
Molecular Formula C7H13NS
Molecular Weight (g/mol) 143.248
MDL Number MFCD01680822
InChI Key CSACPVARWHDAET-UHFFFAOYSA-N
Synonym 2-ethyl-4,5-dimethyl-2,5-dihydrothiazole, 4,5-dimethyl-2-ethyl-3-thiazoline, 2-ethyl-2,5-dihydro-4,5-dimethylthiazole, 2-ethyl-4,5-dimethyl-3-thiazoline, 2-ethyl-4,5-dimethyl thiazoline, fema no. 3620, 3-thiazoline, 2-ethyl-4,5-dimethyl, 3-thiazoline, 4,5-dimethyl-2-ethyl, thiazole, 2,5-dihydro-4,5-dimethyl-2-ethyl, thiazole, 2-ethyl-2,5-dihydro-4,5-dimethyl
PubChem CID 5362584
IUPAC Name 2-ethyl-4,5-dimethyl-2,5-dihydro-1,3-thiazole
SMILES CCC1N=C(C(S1)C)C

Specifications

Density 0.981
Boiling Point 65°C (3 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.4955
Quantity 10 g
Formula Weight 143.26
Percent Purity 99%
Chemical Name or Material 2-Ethyl-4,5-dimethyl-3-thiazoline, Cis + Trans
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.