CAS RN 768-91-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Methyl Adamantane, TRC
CAS: 768-91-2 Molecular Formula: C11 H18 Molecular Weight (g/mol): 150.26 Synonym: 1-Methyl-tricyclo[3.3.1.13,7]decane,1-Methyl-adamantane IUPAC Name: 1-methyladamantane SMILES: C[C@@]12C[C@@H]3C[C@@H](C[C@@H](C3)C1)C2