accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2

CAS RN 768-94-5

IUPAC Name: adamantan-1-amine
Synonyms: amantadine aminoadamantane viregyt amantadina virosol amantadinum amantidine
Molecular Weight (g/mol): 151.25
Molecular Formula: C10H17N
InChi Key: DKNWSYNQZKUICI-UHFFFAOYSA-N
SMILES: NC12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)