CAS RN 76824-16-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]propanamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]propanamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]propanamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.