Learn More
3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]propanamide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-[((2-[(Diaminomethylene)-amino]thiazol-4-yl)methyl)sulfanyl]-propanamide |
| CAS | 76824-16-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8 H13 N5 O S2 |
| Quantity | 100 mg |
| Synonym | Propanamide, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]- (9CI), 3-[[[2-[(Diaminomethylene)-amino]thiazol-4-yl]methyl]sulfanyl]-propanamide, Famotidine Imp. D (EP) |
| InChI Formula | InChI=1S/C8H13N5OS2/c9-6(14)1-2-15-3-5-4-16-8(12-5)13-7(10)11/h4H,1-3H2,(H2,9,14)(H4,10,11,12,13) |
| SMILES | NC(=Nc1nc(CSCCC(=O)N)cs1)N |
| IUPAC Name | 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]propanamide |
| Molecular Weight (g/mol) | 259.35 |
| Show More |
Product Title
¿Detecta una oportunidad de mejora?