CAS RN 76939-46-3
CAS RN 76939-46-3
IUPAC Name:
sodium 6-(6-amino-8-bromo-9H-purin-9-yl)-2,7-dihydroxy-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-one
Synonyms:
sodium 6-(6-amino-8-bromopurin-9-yl)-2,7-dihydroxy-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-one
Molecular Weight (g/mol):
431.09
Molecular Formula:
C10H11BrN5NaO6P
InChi Key:
DMRMZQATXPQOTP-UHFFFAOYNA-N
SMILES:
[Na+].NC1=C2N=C(Br)N(C3OC4COP(O)(=O)OC4C3O)C2=NC=N1
1
–
4
of
4
results
1
8-Bromo-cAMP Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Promotions Available
Tocris Bioscience™ 8-Bromo-cAMP, sodium salt
Tocris Bioscience™ 8-Bromo-cAMP, sodium salt
CAS: 76939-46-3 Molecular Formula: C10H11BrN5NaO6P
3
8-Bromo-cAMP Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
8-Bromo-cAMP Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.