missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 8-Bromo-cAMP, sodium salt

Product Code. 30001206
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001206 10 mg 10mg
18055122 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001206 Supplier Tocris Bioscience™ Supplier No. 1140/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell-permeable cAMP analog

Cell-permeable cAMP analog; activator of protein kinase A.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76939-46-3
Molecular Formula C10H11BrN5NaO6P

Specifications

Quantity 10 mg
Formula Weight 430.09
Purity >99%
Molecular Weight (g/mol) 431.09
Chemical Name or Material 8-Bromo-cAMP, sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.