accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

1HCl

CAS RN 7698-05-7

IUPAC Name: proton chloride
Synonyms: proton chloride
Molecular Weight (g/mol): 36.46
Molecular Formula: ClH
InChi Key: VEXZGXHMUGYJMC-FTGQXOHASA-N
SMILES: [1H+].[Cl-]
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Grade 
  • (4)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)