accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 77-85-0

CH3HOOHHO

CAS RN 77-85-0

IUPAC Name: 2-(hydroxymethyl)-2-methylpropane-1,3-diol
Synonyms: trimet
Molecular Weight (g/mol): 120.15
Molecular Formula: C5H12O3
InChi Key: QXJQHYBHAIHNGG-UHFFFAOYSA-N
SMILES: CC(CO)(CO)CO
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)