accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 77-90-7

H3COOOOCH3OCH3OOOCH3

CAS RN 77-90-7

IUPAC Name: 1,2,3-tributyl 2-(acetyloxy)propane-1,2,3-tricarboxylate
Synonyms: acetyltributylcitrate
Molecular Weight (g/mol): 402.48
Molecular Formula: C20H34O8
InChi Key: QZCLKYGREBVARF-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CC(CC(=O)OCCCC)(OC(C)=O)C(=O)OCCCC
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results