missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyl Acetylcitrate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Tributyl Acetylcitrate
Specifications
Specifications
| Chemical Name or Material | Tributyl acetylcitrate |
| CAS | 77-90-7 |
| Analyte or Component Names | Tributyl acetylcitrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H34 O8 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Tributyl Acetylcitrate, 1,2,3-Propanetricarboxylic acid, 2-(acetyloxy)-, tributyl ester (9 CI), Citric acid, tributyl ester, acetate (6 CI,7 CI,8 CI), 2-Acetoxy-1,2,3-propanetricarboxylic acid tributyl ester, ATBC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC, Kanatol 3400 AC |
| InChI Formula | InChI=1 S/C20H34O8/c1-5-8-11-25-17(22)14-20(28-16(4)21,19(24)27-13-10-7-3)15-18(23)26-12-9-6-2/h5-15H2,1-4H3 |
| SMILES | CCCCOC(=O)CC(CC(=O)OCCCC)(OC(=O)C)C(=O)OCCCC |
| IUPAC Name | tributyl 2-acetyloxypropane-1,2,3-tricarboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction