accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOHOOCH3OOCH3

CAS RN 77-94-1

IUPAC Name: 1,2,3-tributyl 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: tributyl citrate
Molecular Weight (g/mol): 360.45
Molecular Formula: C18H32O7
InChi Key: ZFOZVQLOBQUTQQ-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results