missing translation for 'onlineSavingsMsg'
Learn More

Tributyl Citrate, TRC

Product Code. 30449219
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449219 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449219 Supplier Toronto Research Chemicals Supplier No. TRCT773395500G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tributyl citrate
CAS 77-94-1
Molecular Formula C18 H32 O7
Quantity 500 g
Synonym Tributyl 2-Hydroxypropane-1,2,3-tricarboxylate, Tributyl Acetylcitrate Imp. A (EP), Tributyl Citrate, 1,2,3-Propanetricarboxylic acid','2-hydroxy-','tributyl ester (9CI), Citric acid','tributyl ester (6CI,7CI,8CI), 2-Hydroxy-1,2,3-propanetricarboxylic acid tributyl ester, Butyl citrate, Citroflex 4, Citroflex C 4, Citrofol B 1, Citrofol BI, Morflex TBC, NSC 8491, Scandinol SP 21, TBC, Tri-n-butyl citrate, Tributyl citrate, Uniplex 83
InChI Formula InChI=1S/C18H32O7/c1-4-7-10-23-15(19)13-18(22,17(21)25-12-9-6-3)14-16(20)24-11-8-5-2/h22H,4-14H2,1-3H3
SMILES CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC
IUPAC Name tributyl 2-hydroxypropane-1,2,3-tricarboxylate
Molecular Weight (g/mol) 360.44
Formula Weight 360.2148
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.