accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

KKOSOOOOSOOO

CAS RN 7727-21-1

IUPAC Name: dipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
Synonyms: potassium persulfate
Molecular Weight (g/mol): 270.31
Molecular Formula: K2O8S2
InChi Key: USHAGKDGDHPEEY-UHFFFAOYSA-L
SMILES: [K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (7)
  • (19)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (7)
Physical Form 
  • (7)
  • (5)
  • (2)
  • (6)
Percent Purity 
  • (10)
  • (3)
  • (2)
  • (3)
  • (1)
Grade 
  • (4)
  • (5)
  • (5)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (5)
  • (3)
  • (7)
  • (9)
  • (2)
  • (7)
  • (19)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (7)
  • (7)
  • (5)
  • (2)
  • (6)
  • (10)
  • (3)
  • (2)
  • (3)
  • (1)
  • (4)
  • (5)
  • (5)
  • (1)
  • (3)