missing translation for 'onlineSavingsMsg'
Learn More

Potassium Persulfate, TRC

Product Code. 30422928
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422928 1 g -
30412595 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30422928 Supplier Toronto Research Chemicals Supplier No. TRCP6982201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-21-1
Molecular Formula K2O8S2
Molecular Weight (g/mol) 270.32
Synonym Anthion, Dipotassium peroxodisulfate, Dipotassium peroxydisulfate, Dipotassium persulfate, F 210 Hygisept, Potassium dipersulfate, Potassium peroxydisulfate, Potassium peroxydisulfate (K2(S2O8)), Potassium Peroxydisulphate, Potassium persulfate, Virkon S
IUPAC Name dipotassium;sulfonatooxy sulfate
SMILES [K+].[K+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/2K.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2
Formula Weight 269.8309
Chemical Name or Material Potassium Persulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.