accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOONFFF

CAS RN 773108-67-1

IUPAC Name: 4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Synonyms: 4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Molecular Weight (g/mol): 283.21
Molecular Formula: C13H8F3NO3
InChi Key: CCNGYKFALZVXHZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OC2=NC=C(C=C2)C(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results