CAS RN 773871-49-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
6-Chloro-4-(2-chlorophenyl)-2-quinazolinemethanol, TRC
CAS: 773871-49-1 Molecular Formula: C15 H10 Cl2 N2 O Molecular Weight (g/mol): 305.16 Synonym: 6-Chloro-4-(2-chlorophenyl)-2-quinazoline-methanol IUPAC Name: [6-chloro-4-(2-chlorophenyl)quinazolin-2-yl]methanol SMILES: OCc1nc(c2ccccc2Cl)c3cc(Cl)ccc3n1
2
6-Chloro-4-(2-chlorophenyl)-2-quinazoline-methanol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.