Learn More
6-Chloro-4-(2-chlorophenyl)-2-quinazolinemethanol, TRC
Product Code. 30010450
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
100 mg
missing translation for 'unitSize':
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30010450 100 mg 100mg
30003943 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Product Code. 30010450 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC364815100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 773871-49-1
Molecular Formula C15 H10 Cl2 N2 O
Molecular Weight (g/mol) 305.16
Synonym 6-Chloro-4-(2-chlorophenyl)-2-quinazoline-methanol
IUPAC Name [6-chloro-4-(2-chlorophenyl)quinazolin-2-yl]methanol
SMILES OCc1nc(c2ccccc2Cl)c3cc(Cl)ccc3n1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C15H10Cl2N2O/c16-9-5-6-13-11(7-9)15(19-14(8-20)18-13)10-3-1-2-4-12(10)17/h1-7,20H,8H2
Formula Weight 304.017
Percent Purity >95%
Chemical Name or Material 6-Chloro-4-(2-chlorophenyl)-2-quinazoline-methanol
Grade HPLC