accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7757-93-9

CaOHPOOO

CAS RN 7757-93-9

IUPAC Name: calcium hydrogen phosphate
Synonyms: calcium hydrogen phosphate calcium phosphate dibasic anhydrous phosphoric acid, calcium salt (1:1) secondary calcium phosphate CaHPO4 monocalcium acid phosphate
Molecular Weight (g/mol): 136.06
Molecular Formula: CaHO4P
InChi Key: FUFJGUQYACFECW-UHFFFAOYSA-L
SMILES: [Ca++].OP([O-])([O-])=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (3)
Grade 
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (7)