accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClCu

CAS RN 7758-89-6

IUPAC Name: λ¹-copper(1+) chloride
Synonyms: copper(I) chloride
Molecular Weight (g/mol): 99
Molecular Formula: ClCu
InChi Key: OXBLHERUFWYNTN-UHFFFAOYSA-M
SMILES: [Cl-].[Cu+]
Molecular Weight (g/mol) 
  • (3)
  • (16)
  • (17)
Quantity 
  • (3)
  • (1)
  • (8)
  • (2)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (8)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (7)
  • (3)
  • (2)
Physical Form 
  • (8)
  • (3)
  • (14)
Grade 
  • (4)
  • (1)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (1)
  • (14)
  • (7)
  • (3)
  • (3)
  • (16)
  • (17)
  • (3)
  • (1)
  • (8)
  • (2)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (8)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (7)
  • (3)
  • (2)
  • (8)
  • (3)
  • (14)
  • (4)
  • (1)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)