accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaOClOO

CAS RN 7775-09-9

IUPAC Name: sodium chlorate
Synonyms: sodium chlorate
Molecular Weight (g/mol): 106.44
Molecular Formula: ClNaO3
InChi Key: YZHUMGUJCQRKBT-UHFFFAOYSA-M
SMILES: [Na+].[O-][Cl](=O)=O
Quantity 
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
Physical Form 
  • (3)
  • (3)
  • (5)
Percent Purity 
  • (9)
  • (1)
Grade 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (1)
  • (6)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (3)
  • (5)
  • (9)
  • (1)
  • (3)
  • (1)
  • (3)