accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3NFBr

CAS RN 77771-03-0

IUPAC Name: (3-bromo-4-fluorophenyl)methanaminium
Synonyms: (3-bromo-4-fluorophenyl)methanaminium
Molecular Weight (g/mol): 205.05
Molecular Formula: C7H8BrFN
InChi Key: WIRTWOIWFFCEPB-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC=C(F)C(Br)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results