missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzyl alcohol, 97%

Product Code. 15483957
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483957 25 g 25g
15473957 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483957 Supplier Thermo Scientific Alfa Aesar Supplier No. H30604.14
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 77771-03-0 | C7H6BrFO | 205.026 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77771-03-0
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.026
MDL Number MFCD00143093
InChI Key HNVVROFWONXXGO-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorobenzyl alcohol, 3-bromo-4-fluorophenyl methanol, 3-bromo-4-fluorobenzylalcohol, 3-bromo-4-fluoro-phenyl-methanol, 3-bromo-4-fluorophenyl methan-1-ol, pubchem4884, 4-fluoro-3-bromobenzyl alcohol, 3-bromo-4-fluoro-benzyl alcohol, 4-fluoro-3-bromo-benzyl alcohol, 3-bromo-4-fluoro benzyl alcohol
PubChem CID 2773351
IUPAC Name (3-bromo-4-fluorophenyl)methanol
SMILES C1=CC(=C(C=C1CO)Br)F

Specifications

Boiling Point 80°C (0.5 mmHg)
Quantity 25 g
Formula Weight 205.03
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-fluorobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.