accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaNaOHPOOOH2O7

CAS RN 7782-85-6

IUPAC Name: disodium phosphoric acid heptahydrate
Synonyms: disodium phosphoric acid heptahydrate
Molecular Weight (g/mol): 270.08
Molecular Formula: H17Na2O11P
InChi Key: PYLIXCKOHOHGKQ-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O
Quantity 
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (12)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (22)
Percent Purity 
  • (6)
  • (3)
  • (4)
  • (1)
  • (1)
  • (5)
Physical Form 
  • (3)
  • (1)
  • (10)
  • (1)
  • (3)
Grade 
  • (7)
  • (3)
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (6)
  • (3)
  • (1)
  • (6)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (12)
  • (1)
  • (3)
  • (22)
  • (6)
  • (3)
  • (4)
  • (1)
  • (1)
  • (5)
  • (3)
  • (1)
  • (10)
  • (1)
  • (3)
  • (7)
  • (3)
  • (3)
  • (2)
  • (2)