missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Heptahydrate, TRC

Product Code. 30428445
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428445 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428445 Supplier Toronto Research Chemicals Supplier No. TRCS667035100G
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate
CAS 7782-85-6
Quantity 100 g
Molecular Formula HNa2O4P • 7(H2O)
Synonym Disodium Hydrogen Phosphate Heptahydrate, Disodium Orthophosphate Heptahydrate, Disodium Phosphate Heptahydrate, Sodium Monohydrogen Phosphate Heptahydrate, Sodium Phosphate (Na2HPO4) Heptahydrate
InChI Formula InChI=1S/2Na.H3O4P.7H2O/c;;1-5(2,3)4;;;;;;;/h;;(H3,1,2,3,4);7*1H2/q2*+1;;;;;;;;/p-2
SMILES O.O.O.O.O.O.O.O=P([O-])([O-])O.[Na+].[Na+]
IUPAC Name disodium;hydrogen phosphate;heptahydrate
Molecular Weight (g/mol) 141.9671802
Formula Weight 268.0147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.