accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7787-69-1

IUPAC Name: caesium(1+) bromide
Synonyms: caesium bromide
Molecular Weight (g/mol): 212.81
Molecular Formula: BrCs
InChi Key: LYQFWZFBNBDLEO-UHFFFAOYSA-M
SMILES: [Br-].[Cs+]
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
Physical Form 
  • (2)
  • (4)
  • (2)
Percent Purity 
  • (4)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (8)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)