CAS RN 7787-70-4
Copper(I) bromide, 98%, extra pure
CAS: 7787-70-4 Molecular Formula: BrCu Molecular Weight (g/mol): 143.45 MDL Number: MFCD00010969 InChI Key: NKNDPYCGAZPOFS-UHFFFAOYSA-M Synonym: copper i bromide,copper bromide cubr,copper 1+ bromide,copper monobromide,cuprousbromide,hsdb 270,cubr,copper 1 bromide PubChem CID: 24593 IUPAC Name: bromocopper SMILES: [Cu+].[Br-]
Copper(I) bromide, Puratronic™, 99.998% (metals basis)
CAS: 7787-70-4 Molecular Formula: BrCu Molecular Weight (g/mol): 143.45 MDL Number: MFCD00010969 InChI Key: NKNDPYCGAZPOFS-UHFFFAOYSA-M Synonym: copper i bromide,copper bromide cubr,copper 1+ bromide,copper monobromide,cuprousbromide,hsdb 270,cubr,copper 1 bromide PubChem CID: 24593 IUPAC Name: bromocopper SMILES: [Cu+].[Br-]
Copper(I) bromide, 98%
CAS: 7787-70-4 Molecular Formula: BrCu Molecular Weight (g/mol): 143.45 MDL Number: MFCD00010969 InChI Key: NKNDPYCGAZPOFS-UHFFFAOYSA-M Synonym: copper i bromide,copper bromide cubr,copper 1+ bromide,copper monobromide,cuprousbromide,hsdb 270,cubr,copper 1 bromide PubChem CID: 24593 IUPAC Name: bromocopper SMILES: [Cu+].[Br-]
Copper (I) Bromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Copper (I) Bromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Copper (I) Bromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Copper(I) Bromide, 98%, Honeywell™ Fluka™
CAS: 7787-70-4 Molecular Formula: BrCu Molecular Weight (g/mol): 143.45 MDL Number: MFCD00010969 InChI Key: NKNDPYCGAZPOFS-UHFFFAOYSA-M Synonym: copper i bromide,copper bromide cubr,copper 1+ bromide,copper monobromide,cuprousbromide,hsdb 270,cubr,copper 1 bromide PubChem CID: 24593 IUPAC Name: λ¹-copper(1+) bromide SMILES: [Cu+].[Br-]
Copper(I) Bromide, ≥98.0% (RT), Honeywell™
CAS: 7787-70-4 Molecular Formula: BrCu Molecular Weight (g/mol): 143.45 MDL Number: MFCD00010969 InChI Key: NKNDPYCGAZPOFS-UHFFFAOYSA-M Synonym: copper i bromide,copper bromide cubr,copper 1+ bromide,copper monobromide,cuprousbromide,hsdb 270,cubr,copper 1 bromide PubChem CID: 24593 SMILES: [Cu+].[Br-]