missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Bromide, 98%, Honeywell™ Fluka™

Product Code. 15631950 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass Bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15631950 50 g Glass Bottle 50g
15621950 250 g Glass Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15631950 Supplier Honeywell-Fluka Supplier No. 21286550G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7787-70-4
Molecular Formula BrCu
Molecular Weight (g/mol) 143.45
MDL Number MFCD00010969
InChI Key NKNDPYCGAZPOFS-UHFFFAOYSA-M
Synonym copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide
PubChem CID 24593
IUPAC Name λ¹-copper(1+) bromide
SMILES [Cu+].[Br-]

Specifications

Boiling Point 1345°C
Melting Point 492°C
Color Blue Green
Physical Form Solid
Quantity 50 g
Linear Formula CuBr
UN Number NONH for all modes of transport
Formula Weight 143.45g/mol
Percent Purity 98%
Packaging Glass Bottle
Density 4.71g/mL at 25°C
Chemical Name or Material Copper(I) Bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.