accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7790-29-6

RbI

CAS RN 7790-29-6

IUPAC Name: rubidium(1+) iodide
Synonyms: rubidium iodide
Molecular Weight (g/mol): 212.37
Molecular Formula: IRb
InChi Key: WFUBYPSJBBQSOU-UHFFFAOYSA-M
SMILES: [Rb+].[I-]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results