CAS RN 7790-78-5
IUPAC Name:
cadmium(2+) dichloride
Synonyms:
cadmium chloride
Molecular Weight (g/mol):
183.31
Molecular Formula:
CdCl2
InChi Key:
YKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES:
[Cl-].[Cl-].[Cd++]
1
–
5
of
5
results
1
Cadmium chloride hemipentahydrate, 99+%
CAS: 7790-78-5 Molecular Formula: 2·5 H2O Molecular Weight (g/mol): 228.34 MDL Number: MFCD00799071
2
Cadmium chloride hemipentahydrate, ACS reagent
CAS: 7790-78-5 Molecular Formula: 2·5 H2O Molecular Weight (g/mol): 228.34 MDL Number: MFCD00799071
3
Cadmium chloride hemipentahydrate, ACS, 79.5-81.0%
CAS: 7790-78-5 MDL Number: MFCD00799071
5
Cadmium Chloride Hemi(Pentahydrate), 79.5 to 81.0%, ACS Reagent, Solstice
CAS: 7790-78-5 MDL Number: MFCD00799071