accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 78-07-9

IUPAC Name: triethoxy(ethyl)silane
Synonyms: ethyl triethoxysilane
Molecular Weight (g/mol): 192.33
Molecular Formula: C8H20O3Si
InChi Key: DENFJSAFJTVPJR-UHFFFAOYSA-N
SMILES: CCO[Si](CC)(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results