missing translation for 'onlineSavingsMsg'
Learn More

Ethyltriethoxysilane, 96%

Product Code. 11349205
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349205 100 mL 100mL
11339205 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349205 Supplier Thermo Scientific Alfa Aesar Supplier No. L16495.AE
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78-07-9 | C8H20O3Si | 192.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-07-9
Molecular Formula C8H20O3Si
Molecular Weight (g/mol) 192.33
MDL Number MFCD00026754
InChI Key DENFJSAFJTVPJR-UHFFFAOYSA-N
Synonym ethyltriethoxysilane, triethoxy ethyl silane, silane, triethoxyethyl, a 15 silane, union carbide a-15, triethoxy-ethylsilane, silane, ethyltriethoxy, silane, ethyl triethoxy a-15, ethaneorthosiliconic acid, triethyl ester, ethyl triethoxysilane
PubChem CID 6515
IUPAC Name triethoxy(ethyl)silane
SMILES CCO[Si](CC)(OCC)OCC

Specifications

Boiling Point 158°C to 159°C
Melting Point -78°C
Quantity 100 mL
UN Number UN1993
Beilstein 1362028
Flash Point 40°C (104°F)
Formula Weight 192.33
Percent Purity 96%
Refractive Index 1.392
Sensitivity Moisture sensitive
Density 0.896
Chemical Name or Material Ethyltriethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.